Learn More
Sino Biological Anti-GDNF Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 80050RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat GDNF (rR GDNF; Catalog#80050-R07B; Q07731-1; Ser78-Ile211). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-2849