Learn More
Sino Biological Anti-GFR Alpha-1/GFRA1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 80021RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat GFRA1 (Catalog#80021-R08H; Q62997-1; Met1-Leu445). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-2868