Learn More
Sino Biological Anti-GFR Alpha-1/GFRA1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 80021RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat GFRA1 (rR GFRA1; Catalog#80021-R08H; Q62997-1; Met 1-Leu 445). GFRA1 specific IgG was purified by Rat GFRA1 affinity chromatography.
Catalog No. 50-255-2867