Learn More
Sino Biological Rat GGT1 Protein 100ug (Sino Biological)
Supplier:  Sino Biological 80364R07H100
Encompass_Preferred
A DNA sequence encoding the rat GGT1 (P07314) (Pro27-Tyr568) was expressed, fused with a polyhistidine tag at the N-terminus.
Catalog No. 50-237-0633