Learn More
Sino Biological Anti-Growth Hormone Receptor Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 80029RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat Growth Hormone Receptor / GHR / GHBP (Catalog#80029-R08H; P16310-1; Met1-Arg265). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-2865