Learn More
Sino Biological Anti-Growth Hormone Receptor Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 80029RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat Growth Hormone Receptor / GHR / GHBP (rR Growth Hormone Receptor / GHR / GHBP; Catalog#80029-R08H; P16310-1; Met 1-Arg 265). Growth Hormone Receptor / GHR / GHBP specific IgG was purified by Rat Growth Hormone Receptor / GHR / GHBP affinity chromatography.
Catalog No. 50-255-2864