Learn More
Biomatik Corporation Rat GLI Family Zinc Finger Protein 1 (GLI1) ELISA Kit
Supplier:  Biomatik Corporation EKU10866
Encompass_Preferred
Rat GLI Family Zinc Finger Protein 1 (GLI1) ELISA Kit for detecting GLI1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.113ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: G3V6X8. Gene ID: 140589. Target Synonyms: Glioma-Associated Oncogene Family Zinc Finger 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8276