Learn More
American Research Products Inc Rat glial cell line deriVed ne
Catalog No. 5000326321
Encompass
Rat glial cell line-deriVed neurotrophic factor GDNF ELISA KIT from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 81 pg/mL Sample serum plasma cell culture supernates Method Sandwich
Catalog No. 50-003-26321 Supplier American Research Products Inc Supplier No. CSBE04566R
Rat glial cell line-deriVed neurotrophic factor GDNF ELISA KIT from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 81 pg/mL Sample serum plasma cell culture supernates Method Sandwich