Learn More
Biomatik Corporation Rat Glial Cell Line Derived Neurotrophic Factor (GDNF) CLIA Kit
Supplier:  Biomatik Corporation EKU09754
Encompass_Preferred
Rat Glial Cell Line Derived Neurotrophic Factor (GDNF) CLIA Kit for detecting GDNF in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 6.86-5,000pg/ml. Sensitivity: 2.99pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q07731. Gene ID: 25453. Target Synonyms: ATF1; ATF2; HFB1-GDNF; Astrocyte-Derived Trophic Factor; Glial Derived Neurotrophic Factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8445