Learn More
Biotang Inc Rat Glucagon-like Peptide 1,GLP-1 ELISA Kit, 96T
Supplier: Biotang Inc R6598
Rat Glucagon-like Peptide 1,GLP-1 ELISA Kit, 96T. Lead time of this kit is 1-2 weeks. Shipped by FedEx air. Storage: 2 - 8 C. Shelf Time: 6 months. Kit Experimental Time:< 2 hs. TSZELISA brand.