Learn More
Crystal Chem Inc Rat Glucagon ELISA Kit, 96-wells, Uses 10uL of sample
SDP
Supplier:  Crystal Chem Inc 81519
Encompass
The Rat Glucagon ELISA is used to quantify levels of rat glucagon in plasma, serum, and cell culture using only a 10 µL sample volume. Highlights: Kits use a sample volume of only 10 µL. High sensitivity of 1.1 pg/mL. Dynamic Range of 7.8 - 500 pg/mL. Designed for rat samples.Crystal Chem is a worldwide leading supplier of mouse and rat ELISA kits for research use. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7904