Learn More
Biomatik Corporation Rat Glucagon Like Peptide 1 (GLP1) ELISA Kit
Supplier:  Biomatik Corporation EKU12295
Encompass_Preferred
Rat Glucagon Like Peptide 1 (GLP1) ELISA Kit for detecting GLP1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 12.35-1,000pg/ml. Sensitivity: 4.94pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P06883. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-7989