Learn More
Biomatik Corporation Rat Glucagon Like Peptide 2 (GLP2) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU12012
Encompass_Preferred
Rat Glucagon Like Peptide 2 (GLP2) Mini sample ELISA Kit for detecting GLP2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 123.5-10,000pg/ml. Sensitivity: 43.4pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 3h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7290