Learn More
American Research Products Inc Rat glucocorticoid receptor G
Catalog No. 5000326770
Encompass
Rat glucocorticoid receptor GR ELISA Kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-26770 Supplier American Research Products Inc Supplier No. CSBE08747R
Rat glucocorticoid receptor GR ELISA Kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich