Learn More
Cayman Chemical Rat GlucocortIcoId RECPTR R 96
SDP
Catalog No. 502947525
Encompass_Preferred
rGR Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments. [INDIGO Catalog Nos. R00201, R00201-32]
Catalog No. 50-294-7525 Supplier Cayman Chemical Supplier No. 3351096 WELL
rGR Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments. [INDIGO Catalog Nos. R00201, R00201-32]