Learn More
Crystal Chem Inc Rat Glucose Assay Kit, 10-Kit Pack, 96-tests per well
SDP
Catalog No. 501947910
Encompass_Preferred
The Rat Glucose 10-kit pack is an economical option for high-volume users of our Rat Glucose Assay Kit offered as ten individual kits.Highlights: Easy-to-use, results in 5 minutes. Kits use a sample size of only 2 µL. For use with serum, plasma, and urine. Designed for rat samples. Crystal Chem is a worldwide leading supplier of mouse and rat ELISA kits for research use. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7910 Supplier Crystal Chem Inc Supplier No. 81695
The Rat Glucose 10-kit pack is an economical option for high-volume users of our Rat Glucose Assay Kit offered as ten individual kits.Highlights: Easy-to-use, results in 5 minutes. Kits use a sample size of only 2 µL. For use with serum, plasma, and urine. Designed for rat samples. Crystal Chem is a worldwide leading supplier of mouse and rat ELISA kits for research use. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.