Learn More
Crystal Chem Inc Rat Glucose Assay Kit, 96-tests
SDP
Catalog No. 501947908
Encompass_Preferred
The Rat Glucose Assay is used to quantify levels of glucose in rat serum, plasma, and urine using only a 2 µL sample volume. Highlights: Easy-to-use, results in 5 minutes. Kits use a sample size of only 2 µL. For use with serum, plasma, and urine. Designed for rat samples. Crystal Chem is a worldwide leading supplier of mouse and rat ELISA kits for research use. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7908 Supplier Crystal Chem Inc Supplier No. 81693
The Rat Glucose Assay is used to quantify levels of glucose in rat serum, plasma, and urine using only a 2 µL sample volume. Highlights: Easy-to-use, results in 5 minutes. Kits use a sample size of only 2 µL. For use with serum, plasma, and urine. Designed for rat samples. Crystal Chem is a worldwide leading supplier of mouse and rat ELISA kits for research use. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.