Learn More
Biomatik Corporation Rat Glutamate Decarboxylase 1, Brain (GAD1) ELISA Kit
Supplier:  Biomatik Corporation EKU09141
Encompass_Preferred
Rat Glutamate Decarboxylase 1, Brain (GAD1) ELISA Kit for detecting GAD1 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P18088. Gene ID: 24379. Target Synonyms: GAD67; SCP; 67 kDa glutamic acid decarboxylase; Glutamate decarboxylase 67 kDa isoform. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7411