Learn More
Biomatik Corporation Rat Glutamate Receptor, Ionotropic, N-Methyl-D-Aspartate 2A (GRIN2A) ELISA Kit
Supplier:  Biomatik Corporation EKU09208
Encompass_Preferred
Rat Glutamate Receptor, Ionotropic, N-Methyl-D-Aspartate 2A (GRIN2A) ELISA Kit for detecting GRIN2A in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.78-50ng/ml. Sensitivity: 0.30ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q00959. Gene ID: 24409. Target Synonyms: NMDAR2A; NR2A; Glutamate [NMDA] receptor subunit epsilon-1; N-methyl D-aspartate receptor subtype 2A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5007