Learn More
American Research Products Inc Rat glutamic acid decarboXylas
Catalog No. 5000329116
Encompass
Rat glutamic acid decarboXylase (GAD) autoantibody (IgG) ELISA Kit from CUSABIO Detection Range 7 8 pg/mL-500 pg/mL Sensitivity 1 95 pg/mL Sample serum plasma Method Sandwich
Catalog No. 50-003-29116 Supplier American Research Products Inc Supplier No. CSBE08654R
Rat glutamic acid decarboXylase (GAD) autoantibody (IgG) ELISA Kit from CUSABIO Detection Range 7 8 pg/mL-500 pg/mL Sensitivity 1 95 pg/mL Sample serum plasma Method Sandwich