Learn More
Biomatik Corporation Rat Glutamine synthetase (GS) ELISA Kit
Supplier:  Biomatik Corporation EKU04468
Encompass_Preferred
Rat Glutamine synthetase (GS) ELISA Kit for detecting GS in serum, plasma, tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.62-1000pg/ml. Sensitivity: 6.2pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P09606. Gene ID: 24957. Target Synonyms: GLNS; GLUL; Glutamate-Ammonia Ligase; Glutamate decarboxylase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6281