Learn More
Biomatik Corporation Rat Glutathione Peroxidase 4 (GPX4) ELISA Kit
Supplier:  Biomatik Corporation EKU12535
Encompass_Preferred
Rat Glutathione Peroxidase 4 (GPX4) ELISA Kit for detecting GPX4 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.127ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P36970. Gene ID: 29328. Target Synonyms: MCSP; PHGPx; SnGPx; SnPHGPx; Phospholipid Hydroperoxidase; Phospholipid hydroperoxide glutathione peroxidase, mitochondrial. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-7950