Learn More
American Research Products Inc Rat Glutathione PeroXidase GS
Catalog No. 5000329745
Encompass
Rat Glutathione PeroXidase (GSH-PX) ELISA kit from CUSABIO Detection Range 12 5 mIU/mL-800 mIU/mL Sensitivity 3 12 mIU/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29745 Supplier American Research Products Inc Supplier No. CSBE12146R
Rat Glutathione PeroXidase (GSH-PX) ELISA kit from CUSABIO Detection Range 12 5 mIU/mL-800 mIU/mL Sensitivity 3 12 mIU/mL Sample serum plasma tissue homogenates Method Sandwich