Learn More
American Research Products Inc Rat a glutathione S transferas
Catalog No. 5000329228
Encompass
Rat a-glutathione S-transferases a-GST ELISA Kit from CUSABIO Detection Range 0 156 mU/mL-10 mU/mL Sensitivity 0 039 mU/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-29228 Supplier American Research Products Inc Supplier No. CSBE08907R
Rat a-glutathione S-transferases a-GST ELISA Kit from CUSABIO Detection Range 0 156 mU/mL-10 mU/mL Sensitivity 0 039 mU/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich