Learn More
Biomatik Corporation Rat Glutathione S Transferase Alpha 1 (GSTa1) ELISA Kit
Supplier:  Biomatik Corporation EKU04488
Encompass_Preferred
Rat Glutathione S Transferase Alpha 1 (GSTa1) ELISA Kit for detecting GSTa1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.133ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P00502. Gene ID: 24422. Target Synonyms: GSTA1; GST2; GSTA1-1; GTH1; GST class-alpha member 1; GST HA subunit 1; GST-epsilon; Glutathione S-transferase A1, N-terminally processed. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4456