Learn More
Biomatik Corporation Rat Glutathione S Transferase Alpha 3 (GSTa3) ELISA Kit
Supplier:  Biomatik Corporation EKU04490
Encompass_Preferred
Rat Glutathione S Transferase Alpha 3 (GSTa3) ELISA Kit for detecting GSTa3 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.133ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P04904. Gene ID: 24421. Target Synonyms: GST-A3; GSTA3-3; GTA3; GST class-alpha member 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8953