Learn More
Biomatik Corporation Rat Glutathione Synthetase (GSS) ELISA Kit
Supplier:  Biomatik Corporation EKU04502
Encompass_Preferred
Rat Glutathione Synthetase (GSS) ELISA Kit for detecting GSS in serum, plasma, tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.781-50ng/ml. Sensitivity: 0.33ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P46413. Gene ID: 25458. Target Synonyms: Glutathione synthase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5354