Learn More
American Research Products Inc Rat Glycated hemoglobin A1c G
Catalog No. 5000328666
Encompass
Rat Glycated hemoglobin A1c GHbA1c ELISA Kit from CUSABIO Detection Range 0 019 ng/mL-80 ng/mL Sensitivity 0 019 ng/mL Sample lysates for RBC Method Competitive
Catalog No. 50-003-28666 Supplier American Research Products Inc Supplier No. CSBE08140R
Rat Glycated hemoglobin A1c GHbA1c ELISA Kit from CUSABIO Detection Range 0 019 ng/mL-80 ng/mL Sensitivity 0 019 ng/mL Sample lysates for RBC Method Competitive