Learn More
Biomatik Corporation Rat Glycogen Phosphorylase, Liver (PYGL) ELISA Kit
Supplier:  Biomatik Corporation EKU04517
Encompass_Preferred
Rat Glycogen Phosphorylase, Liver (PYGL) ELISA Kit for detecting PYGL in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.56ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P09811. Gene ID: 64035. Target Synonyms: GPLL; GPBB; Hers Disease; Glycogen Storage Disease Type VI. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6877