Learn More
Biomatik Corporation Rat Glycoprotein 130 (gp130) ELISA Kit
Supplier:  Biomatik Corporation EKU04405
Encompass_Preferred
Rat Glycoprotein 130 (gp130) ELISA Kit for detecting gp130 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.625-40ng/ml. Sensitivity: 0.244ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P40190. Target Synonyms: CDw130; CD130; IL6ST; GP130-RAPS; IR6RB; IL6R-Beta; IR6RB; IR6-RB; Interleukin 6 Signal Transducer; Oncostatin M Receptor; Interleukin-6 Receptor Subunit Beta. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6013