Learn More
Biomatik Corporation Rat Glycoprotein 2, Zymogen Granule Membrane (GP2) ELISA Kit
Supplier:  Biomatik Corporation EKU04526
Encompass_Preferred
Rat Glycoprotein 2, Zymogen Granule Membrane (GP2) ELISA Kit for detecting GP2 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.122ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P19218. Target Synonyms: ZAP75; Pancreatic Secretory Granule Membrane Major Glycoprotein GP2; Pancreatic zymogen granule membrane protein GP-2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6348