Learn More
Biomatik Corporation Rat Glypican 4 (GPC4) ELISA Kit
Supplier:  Biomatik Corporation EKU09040
Encompass_Preferred
Rat Glypican 4 (GPC4) ELISA Kit for detecting GPC4 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.1pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q642B0. Gene ID: 317322. Target Synonyms: K-Glypican; Glypican Proteoglycan 4; Secreted glypican-4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4629