Learn More
Biomatik Corporation Rat Golgi Phosphoprotein 2 (GOLPH2) ELISA Kit
Supplier:  Biomatik Corporation EKU10880
Encompass_Preferred
Rat Golgi Phosphoprotein 2 (GOLPH2) ELISA Kit for detecting GOLPH2 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.78-50ng/ml. Sensitivity: 0.31ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: D4AEL2. Target Synonyms: GOLM1; GP73; C9orf155; Golgi Membrane Protein 1; Golgi Protein 73. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6742