Learn More
BIOHIPPO Rat GP39 Microsample
SDP
Catalog No. 5000314974
Encompass
Rat GP39 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of GP39 in Serum plasma and other biological fluids It provides a detection range of 3 13-200 ng/mL with a sensitivity of 1 26 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat GP39 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat GP39 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat GP39 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14974 Supplier BIOHIPPO Supplier No. BHE15210949
Rat GP39 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of GP39 in Serum plasma and other biological fluids It provides a detection range of 3 13-200 ng/mL with a sensitivity of 1 26 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat GP39 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat GP39 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat GP39 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color