Learn More
American Research Products Inc Rat Granulocyte Colony Stimula
Catalog No. 5000326798
Encompass
Rat Granulocyte Colony Stimulating Factor G-CSF ELISA Kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 81 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-26798 Supplier American Research Products Inc Supplier No. CSBE07340R
Rat Granulocyte Colony Stimulating Factor G-CSF ELISA Kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 81 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich