Learn More
American Research Products Inc Rat Granulocyte Macrophage Col
Catalog No. 5000329906
Encompass
Rat Granulocyte-Macrophage Colony Stimulating Factor GM-CSF ELISA Kit from CUSABIO Detection Range 15 6 pg/ml - 1000 pg/ml Sensitivity 3 9 pg/ml Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29906 Supplier American Research Products Inc Supplier No. CSBE04570R
Rat Granulocyte-Macrophage Colony Stimulating Factor GM-CSF ELISA Kit from CUSABIO Detection Range 15 6 pg/ml - 1000 pg/ml Sensitivity 3 9 pg/ml Sample serum plasma tissue homogenates Method Sandwich