Learn More
Biomatik Corporation Rat Growth Arrest And DNA Damage Inducible Protein Alpha (GADD45a) ELISA Kit
Supplier:  Biomatik Corporation EKU04575
Encompass_Preferred
Rat Growth Arrest And DNA Damage Inducible Protein Alpha (GADD45a) ELISA Kit for detecting GADD45a in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.056ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P48317. Target Synonyms: DDIT1; GADD45; DNA damage-inducible transcript 1 protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5989