Learn More
Biomatik Corporation Rat Growth Differentiation Factor 3 (GDF3) CLIA Kit
Supplier:  Biomatik Corporation EKU09867
Encompass_Preferred
Rat Growth Differentiation Factor 3 (GDF3) CLIA Kit for detecting GDF3 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 5.49-4,000pg/ml. Sensitivity: 2.31pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: A2SY89. Gene ID: 500311. Target Synonyms: Vgr2; Vg-Related Gene 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5649