Learn More
Biomatik Corporation Rat Growth Differentiation Factor 1 (GDF1) CLIA Kit
Supplier:  Biomatik Corporation EKU09870
Encompass_Preferred
Rat Growth Differentiation Factor 1 (GDF1) CLIA Kit for detecting GDF1 in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 2.74-2,000pg/ml. Sensitivity: 0.85pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q1HI69. Gene ID: 306351. Target Synonyms: Embryonic growth/differentiation factor 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5612