Learn More
American Research Products Inc Rat growth differentiation fac
Catalog No. 5000328565
Encompass
Rat growth differentiation factor 15 (GDF15) ELISA Kit from CUSABIO Detection Range 1 56 pg/mL-100 pg/mL Sensitivity 0 39 pg/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-28565 Supplier American Research Products Inc Supplier No. CSBE16317R
Rat growth differentiation factor 15 (GDF15) ELISA Kit from CUSABIO Detection Range 1 56 pg/mL-100 pg/mL Sensitivity 0 39 pg/mL Sample serum plasma tissue homogenates Method Competitive