Learn More
American Research Products Inc Rat Growth differentiation fac
Catalog No. 5000326911
Encompass
Rat Growth/differentiation factor 8 (MSTN) ELISA kit from CUSABIO Detection Range 0 78 ng/mL-50 ng/mL Sensitivity 0 195 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26911 Supplier American Research Products Inc Supplier No. CSBEL015057RA
Rat Growth/differentiation factor 8 (MSTN) ELISA kit from CUSABIO Detection Range 0 78 ng/mL-50 ng/mL Sensitivity 0 195 ng/mL Sample serum plasma tissue homogenates Method Sandwich