Learn More
Biomatik Corporation Rat Growth Differentiation Factor 15 (GDF15) CLIA Kit
Supplier:  Biomatik Corporation EKU09888
Encompass_Preferred
Rat Growth Differentiation Factor 15 (GDF15) CLIA Kit for detecting GDF15 in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 2.74-2,000pg/ml. Sensitivity: 1.21pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q9Z0J6. Gene ID: 29455. Target Synonyms: PDF; MIC1; NAG-1; PLAB; PTGFB; Macrophage inhibitory cytokine 1; NSAID-activated gene 1 protein; Placental bone morphogenetic protein; Prostate differentiation factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4841