Learn More
Raybiotech Inc C-Series Rat Growth Factor Array 1 Kit.
Supplier:  Raybiotech Inc AARGF12
Encompass_Preferred
Detects 14 rat Chemokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: Activin A, BDNF, beta-NGF, bFGF, EGF, Flt-3 Ligand, GDNF, GM-CSF, HGF, IGF-1, PDGF-AA, PDGF-BB, IGFBP-5, VEGF-A
Catalog No. 50-194-8784