Learn More
American Research Products Inc Rat growth hormone releasing h
Catalog No. 5000328965
Encompass
Rat growth hormone releasing hormone GHRH ELISA Kit from CUSABIO Detection Range 3 12 pg/mL-200 pg/mL Sensitivity 0 78 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28965 Supplier American Research Products Inc Supplier No. CSBE06878R
Rat growth hormone releasing hormone GHRH ELISA Kit from CUSABIO Detection Range 3 12 pg/mL-200 pg/mL Sensitivity 0 78 pg/mL Sample serum plasma tissue homogenates Method Sandwich