Learn More
American Research Products Inc Rat growth regulated oncogene
Catalog No. 5000329516
Encompass
Rat growth-regulated oncogene a/melanoma growth stimulating actiVity GROa /MGSA Elisa Kit from CUSABIO Detection Range 7 8pg/mL-500pg/mL Sensitivity 3 9pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-29516 Supplier American Research Products Inc Supplier No. CSBE12997RIS
Rat growth-regulated oncogene a/melanoma growth stimulating actiVity GROa /MGSA Elisa Kit from CUSABIO Detection Range 7 8pg/mL-500pg/mL Sensitivity 3 9pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich