Learn More
American Research Products Inc Rat growth regulated oncogene
Catalog No. 5000329861
Encompass
Rat growth-regulated oncogene a/melanoma growth stimulating actiVity GROa /MGSA Elisa Kit from CUSABIO Detection Range 125 pg/mL-8000 pg/mL Sensitivity 31 25 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29861 Supplier American Research Products Inc Supplier No. CSBE12997R
Rat growth-regulated oncogene a/melanoma growth stimulating actiVity GROa /MGSA Elisa Kit from CUSABIO Detection Range 125 pg/mL-8000 pg/mL Sensitivity 31 25 pg/mL Sample serum plasma tissue homogenates Method Sandwich