Learn More
BIOHIPPO Rat GT Microsample
SDP
Catalog No. 5000314863
Encompass
Rat GT Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of GT in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 12 5-800 pg/mL with a sensitivity of 5 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat GT Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat GT Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat GT biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14863 Supplier BIOHIPPO Supplier No. BHE15210764
Rat GT Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of GT in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 12 5-800 pg/mL with a sensitivity of 5 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat GT Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat GT Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat GT biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color