Learn More
Sino Biological Recombinant Rat GUCY2C/Guanylyl cyclase C Protein (ECD, His Tag)
Supplier:  Sino Biological 84711R08H100
Encompass_Preferred
A DNA sequence encoding the Rat GUCY2C (P23897) (Met1-Tyr560) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-224-6552