Learn More
Sino Biological Rat HAAO Protein 100ug (Sino Biological)
Supplier:  Sino Biological 80627R07E100
Encompass_Preferred
A DNA sequence encoding the rat Haao (NP_064461.1) (Met1-Gly286) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-237-2011