Learn More
BIOHIPPO Rat HAase Microsample
SDP
Catalog No. 5000313711
Encompass
Rat HAase Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of HAase in serum plasma tissue homogenates and other biological fluids It provides a detection range of 3 13-200 ng/mL with a sensitivity of 1 28 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat HAase Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat HAase Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat HAase biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-13711 Supplier BIOHIPPO Supplier No. BHE15212476
Rat HAase Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of HAase in serum plasma tissue homogenates and other biological fluids It provides a detection range of 3 13-200 ng/mL with a sensitivity of 1 28 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat HAase Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat HAase Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat HAase biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color